Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ CCR2, Mouse, Unlabeled, Clone: 8C3.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABF299
Description
C-C chemokine receptor type 2 (UniProt P41597; also known as C-C CKR-2, CC-CKR-2, CCR-2, CCR2, CD192, MCP-1-R, Monocyte chemoattractant protein 1 receptor) is encoded by the CCR2 (also known as CMKBR2) gene (Gene ID 729230) in human. CC chemokine receptors (CCRs) are 7-transmembrane G protein-coupled receptors (GPCRs) that bind CC chemokines (CCLs) and mediate their biological activities. CCRs and CCLs play a central role in physiological and pathological recruitment of immune cells. CCR2 can interact with CCL2, CCL8 and CCL16, it is found on the surface of monocytes, activated memory T cells, B cells, and basophils in humans, and also in peritoneal macrophages in mice. CCR2 plays an important role in the recruitment of monocytes/macrophages and T cells. CCR2-knockout mice show a partial defect in macrophage function, decreased Th1 immune response, reduced delayed-type hypersensitivity reaction, and are more susceptible to pathogenic bacteria infections. Human CCR2 is a 7-transmembrane (a.a. 43-70, 81-100, 115-136, 154-178, 207-226, 244-268, 286-309) GPCR with an N-terminal extracellular domain (a.a. 1-42) and a C-terminoal cytoplasmic domain (a.a. 310-374).
Specifications
CCR2 | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1M Tris-Glycine (pH 7.4) 150mM NaCl with 0.05% Sodium Azide. | |
P41597 | |
GST-tagged recombinant human N-terminal extracellular domain. | |
100 μL | |
Inflammation & Immunology | |
NP_001116513 | |
Human | |
Purified |
Western Blot | |
8C3.1 | |
Unconjugated | |
CCR2, CMKBR2 | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone 8C3.1 targets the N-terminal extracelluar domain of human CCR2 spliced isoforms 1&2 reported by Uniprot (P41597). | |
Stable for 1 year at 2°-8°C from date of receipt. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction