Learn More
Invitrogen™ CCR2 Recombinant Rabbit Monoclonal Antibody (10L1L6)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542780
Description
Positive test controls include: A-549, U-937, Mouse lung, Rat lung. The target is usually found in the following locations: Cell membrane, Multi-pass membrane protein. Immunogen sequence: MLSTSRSRFI RNTNESGEEV TTFFDYDYGA PCHKFDVKQI GAQLLPPLYS LVFIFGFVGN MLVVLILINC KKLKCLTDIY LLNLAISDLL FLITLPLWAH.
CCR2, a chemokine receptor, is a regulator of monocyte/macrophage traffickings and is upregulated in response to inflammation. Resistance to HIV-1 infection or delayed progression to AIDS may be linked to CCR2 polymorphisms. The ligand for CCR2 is monocyte chemoattractant protein (MCP-1), a chemokine produced by endothelial cells, smooth muscle cells, and macrophages. Activation of monocytes by MCP-1 may mediate the monocytic infiltration of tissues in atherosclerosis and other inflammatory disorders including rheumatoid arthritis, alveolitis, and tumor infiltration. Two isoforms are produced by alternative splicing. CCR2 has been reported to be expressed in a wide variety of tissues, including blood, brain, heart, kidney, liver, lung, ovary, pancreas, spinal cord, spleen, thymus, and vessel. Increased expression of CCR2 has been documented in CD3+ lymphocytes in rheumatoid arthritis. ESTs have been isolated from liver/spleen and prostate libraries.
Specifications
CCR2 | |
Recombinant Monoclonal | |
1.5 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O55193, P41597, P51683 | |
Ccr2 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CCR2/CKR2 (P41597). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
10L1L6 | |
Unconjugated | |
Ccr2 | |
C-C chemokine receptor type 2; C-C CHEMOKINE RECEPTOR TYPE 2 (C-C CKR-2) (CC-CKR-2) (CCR-2) (CCR2) (JE/FIC RECEPTOR) (MCP-1 RECEPTOR); C-C CKR-2; C-C motif chemokine receptor 2; Cc-ckr2; CC-CKR-2; Ccr2; CCR-2; Ccr2a; CCR2B; CD192; CD192 antigen; chemoattractant protein-1 receptor; chemokine (C-C motif) receptor 2; chemokine (C-C) receptor 2; chemokine receptor CCR2; Ckr2; Ckr2a; Ckr2b; CMKBR2; hCG_14621; JE/FIC receptor; Mcp1 receptor; MCP-1 receptor; Mcp1r; MCP-1-R; MIP-1 alphaR; mJe-r; Monocyte chemoattractant protein 1 receptor; monocyte chemotactic protein 1 receptor | |
Rabbit | |
Affinity Chromatography | |
RUO | |
12772, 60463, 729230 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.