Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR3 Antibody (5E8-G9-B4), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281046DL594
Description
CCR3 Monoclonal specifically detects CCR3 in Human samples. It is validated for Flow Cytometry.Specifications
CCR3 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
CCR3 | |
Recombinant monoclonal antibody to CCR3 (CD193). | |
0.1 mL | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
1232 | |
Human | |
Purified |
Flow Cytometry | |
5E8-G9-B4 | |
Flow Cytometry | |
b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR-3, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Recognises human CCR3, a chemokine receptor highly expressed of eosinophils, as well as Th2-Tcells, basophils, platelets, mast-cells, mononuclear phagocytes, airway epithelial cells, and hematopoietic progenitor cells (CD34+). | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction