Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR3 Antibody (5E8-G9-B4), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281046F
Description
CCR3 Monoclonal specifically detects CCR3 in Human samples. It is validated for Flow Cytometry.Specifications
| CCR3 | |
| Monoclonal | |
| FITC | |
| PBS | |
| CCR3 | |
| Recombinant monoclonal antibody to CCR3 (CD193). | |
| 0.1 mL | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1232 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 5E8-G9-B4 | |
| Flow Cytometry | |
| b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR-3, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognises human CCR3, a chemokine receptor highly expressed of eosinophils, as well as Th2-Tcells, basophils, platelets, mast-cells, mononuclear phagocytes, airway epithelial cells, and hematopoietic progenitor cells (CD34+). | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction