Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR3 Antibody (83103) [Allophycocyanin], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1551A
Description
CCR3 Monoclonal antibody specifically detects CCR3 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
CCR3 | |
Monoclonal | |
APC | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
83103 | |
Flow Cytometry, CyTOF-ready | |
b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR-3, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841 | |
Y3 rat myeloid cell line transfected with mouse CCR3, Met1-Phe359, Accession # P51678 | |
0.1 mL | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
1232 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction