Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR3 Polyclonal Antibody, Huabio
Rabbit Polyclonal Antibody
Supplier: HUABIO RESEARCH INC HA500279


Description
CCR3 Polyclonal antibody specifically detects CCR3 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
CCR3 | |
Polyclonal | |
Unconjugated | |
CCR3 | |
b-chemokine receptor; CC chemokine receptor 3; C-C chemokine receptor type 3; C-C CKR-3; C-C motif chemokine receptor 3; CC-CKR3; CC-CKR-3; Ccr3; CCR-3; CD193; chemokine (C-C motif) receptor 3; chemokine (C-C) receptor 1,-like 2; chemokine (C-C) receptor 3; CKR3; Cmkbr1l2; CMKBR3; eosinophil CC chemokine receptor 3; eosinophil eotaxin receptor; Macrophage inflammatory protein 1-alpha receptor-like 2; MGC102841; MIP-1 alpha RL2; MIP-1 alphaRL2; probable C-C chemokine receptor type 3 | |
Synthetic peptide within human CCR3 aa 1-34/355. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
P51677 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1232 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction