Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR4 Antibody (12F5), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
| Antigen | CCR4 |
|---|---|
| Clone | 12F5 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
Description
CCR4 Monoclonal antibody specifically detects CCR4 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| CCR4 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5 | |
| Recombinant Human CCR4 protein [UniProt P51679] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 12F5 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Human | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| 1233 | |
| IgG1 | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title