Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR4 Antibody (MM0064-9G12), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212129FR
Description
CCR4 Monoclonal specifically detects CCR4 in Human samples. It is validated for Flow Cytometry.Specifications
CCR4 | |
Monoclonal | |
DyLight 680 | |
P51679 | |
CCR4 | |
Human CCR4 transfected 293 cells | |
0.1 mL | |
Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
1233 | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry (Frozen) | |
MM0064-9G12 | |
Flow Cytometry, Immunohistochemistry-Frozen | |
C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Detects human CCR4, not other CCRs | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction