Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR4 Antibody (MM0064-9G12), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212129MFV500
Description
CCR4 Monoclonal antibody specifically detects CCR4 in Human samples. It is validated for Flow CytometrySpecifications
CCR4 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
MM0064-9G12 | |
Flow Cytometry | |
C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5 | |
Human CCR4 transfected 293 cells | |
0.1 mL | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
1233 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction