Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR4 Antibody, Novus Biologicals™
SDP

Catalog No. NB100721 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100721 0.1 mg
NB017808 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100721 Supplier Novus Biologicals Supplier No. NB100721
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

CCR4 Polyclonal specifically detects CCR4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CCR4
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Flow Cytometry 2 μL/10-6 cells, ELISA 1:25000, Immunohistochemistry 1:250, Immunohistochemistry-Paraffin 1:250
Formulation 10mM KHPO4, 0.14M NaCl and 1.0 mg/ml BSA with 0.1% Sodium Azide
Gene Alias C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5
Gene Symbols CCR4
Host Species Goat
Immunogen Synthetic peptide DTTQDETVYNSYYFYESMP-C corresponds to amino acid residues 8 - 26 of mouse CCR4 with cysteine added for coupling to a carrier protein.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1233
Test Specificity Peptide sequence is < 50 % identical to other mouse chemokine receptors in this region. The antibody recognizes mouse CCR4 and has no cross-reactivity to human CCR4.
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.