Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP186584
Description
CCR4 Polyclonal specifically detects CCR4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CCR4 | |
Polyclonal | |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20-1:50 | |
P51679 | |
CCR4 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LQIYSADTPSSSYTQSTMDHDLHDAL | |
0.1 mL | |
Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
1233 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction