Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR5 Antibody (12D1), PE/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247759PECY55
Description
CCR5 Monoclonal specifically detects CCR5 in Human samples. It is validated for Flow Cytometry.Specifications
CCR5 | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
CCR5 | |
Human native CCR5 protein (Uniprot: P51681) | |
0.1 mL | |
Primary | |
Reacts with the N-terminal extracellular domain of CD195. The CC chemokine receptor 5 (CCR5) is a member of the CC-chemokine receptor family, and has the characteristic structure of a 7 transmembrane G protein-coupled receptor (GPCR). CCR5 regulates trafficking and effector functions of memory/effector Th1 cells, macrophages, NK cells, and immature dendritic cells. CCR5 and its ligands play an important role in viral pathogenesis. CCR5 represents the co-receptor for macrophage (M) and dual (T cell and M)-tropic immunodeficiency viruses. Together with the CD4 binding receptor, CCR5 plays a critical role in HIV entry into the target cells. Moreover, the CCR5 ligands macrophage inflammatory protein (MIP)-1 alpha, MIP-1 beta and RANTES act as endogenous inhibitors of HIV infection, making both CCR5 and its chemokine ligands attractive therapeutic targets for HIV infection. Recent studies have also highlighted the role of CCR5 in a variety of other human diseases, ranging from infectious and inflammatory diseases to cancer. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
12D1 | |
Flow Cytometry | |
C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 | |
Mouse | |
Protein A or G purified | |
Alzheimers Research, Cancer, Chemokine Research, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction | |
1234.0 | |
Human | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction