Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR5 Antibody (45529), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB184I
Description
CCR5 Monoclonal antibody specifically detects CCR5 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
CCR5 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
45529 | |
Flow Cytometry, CyTOF-ready | |
C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 | |
NS0 mouse myeloma cell line transfected with human CCR5, Met1-Leu352, Accession # P51681 | |
0.1 mL | |
Alzheimers Research, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Signal Transduction | |
1234 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction