Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR5 (CD195), Rat anti-Human, Clone: HEK/1/85a/7a, Millipore Sigma™

Catalog No. MABF200525 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF200525 25 μg
MABF2005100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF200525 Supplier MilliporeSigma Supplier No. MABF200525UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-CCR5 (CD195), clone HEK/1/85a/7a, Cat. No. MABF2005, is a rat monoclonal antibody that detects C-C chemokine receptor type 5 (CCR5) and has been tested for use in Flow Cytometry and Immunocytochemistry.

C-C chemokine receptor type 5 (UniProt: P51681; also known as C-C CKR-5, CC-CKR-5, CCR-5, CCR5, CHEMR13, HIV-1 fusion coreceptor, CD195) is encoded by the CCR5 (also known as CMKBR5) gene (Gene ID: 1234) in human. CCR-5 is a multi-pass membrane protein that is expressed in memory T-cells, macrophages, and immature dendritic cells, and is upregulated by proinflammatory cytokines. It is also shown to be highly expressed in spleen, thymus, and on THP-1 and KG-1a cells. Lower expression is observed in ovary and lung. CCR-5 serves as a receptor for a number of inflammatory CC-chemokines and subsequently transduces a signal by increasing intracellular calcium levels. It also plays a role in the control of granulocytic lineage proliferation or differentiation. CCR-5 is acts as a co-receptor (CD4 being the primary receptor) of HIV-1 and HIV-2. It also couples to Gi class of G-proteins to inhibit cAMP production and stimulate calcium release. CCR-5 undergoes sulfation on two tyrosine residues in the N-terminal region, which increases efficiency of HIV-1 entry into cells. It also undergoes phosphorylation at C-terminal serine residues following binding to CC chemokines, especially APO-RANTES. Mutations in CCR5 gene have been linked to insulin-dependent diabetes mellitus. (Ref.: Blanpain, C., et al. (2002). Receptors Channels. 8(1);19-31).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCR5 (CD195)
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone HEK/1/85a/7a
Conjugate Unconjugated
Dilution Immunocytochemistry Analysis: A representative lot detected CCR5 (CD195) in Immunocytochemistry applications (Mueller, A., et. al. (2002). Blood. 99(3):785-91).Flow Cytometry Analysis: A representative lot detected CCR5 (CD195) in Flow Cytometry applications (Mueller, A., et. al. (2002). Blood. 99(3):785-91).
Formulation Purified rat monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias C-C chemokine receptor type 5;C-C CKR-5;CC-CKR-5;CCR-5;CCR5;CHEMR13;HIV-1 fusion coreceptor;CD195
Gene Symbols CCR5;CMKBR5
Host Species Rat
Immunogen CHO cells transfected with full length human CCR-5 (CD195).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_000570.1
Test Specificity Clone HEK/1/85a/7a is a rat monoclonal antibody that specifically detects human C-C-chemokine receptor 5 (CD195).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.