Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR5 Mouse anti-Human, Unconjugated, Clone: 12D1, Abnova™

Catalog No. 89337649 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-649 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-649 Supplier Abnova Corporation Supplier No. MAB21156
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

CCR5 Monoclonal antibody specifically detects CCR5 in Human samples. It is validated for IHC-Fr, IF, Func, Flow Cyt.

Specifications

Antigen CCR5
Applications Immunohistochemistry (Frozen), Immunofluorescence, Functional Assay, Flow Cytometry
Classification Monoclonal
Clone 12D1
Conjugate Unconjugated
Description chemokine (C-C motif) receptor 5
Gene CCR5
Gene Alias CC-CKR-5, CCCKR5, CD195, CKR-5, CKR5, CMKBR5, IDDM22
Host Species Mouse
Immunogen Native purified human CCR5
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1234
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.