Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR5 Rabbit anti-Human, Clone: RC-10, Novus Biologicals™

Rabbit Monoclonal Antibody
$281.00 - $596.50
Specifications
| Antigen | CCR5 |
|---|---|
| Clone | RC-10 |
| Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
CCR5 Monoclonal antibody specifically detects CCR5 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
| CCR5 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 | |
| This antibody was raised by immunizing mice with CCR5. | |
| Primary |
| RC-10 | |
| Monoclonal | |
| Rabbit | |
| Alzheimers Research, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction | |
| PBS | |
| 1234 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title