Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR6 Antibody (140706), Biotin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB590B
Description
CCR6 Monoclonal antibody specifically detects CCR6 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
CCR6 | |
Monoclonal | |
Biotin | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
140706 | |
Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
BN-1, CC-CKR-6, CCR-6, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29C-C CKR-6, GPRCY4DRY-6, lymphocyte, 22 | |
RBL-1 rat basophilic leukemia cell line transfected with mouse CCR6, Met1-Met367, Accession # O54689 | |
0.1 mL | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
1235 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction