Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR6 Antibody (53103R), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB195RMFV500
Description
CCR6 Monoclonal antibody specifically detects CCR6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
| CCR6 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1235 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 53103R | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| BN-1, CC-CKR-6, CCR-6, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29C-C CKR-6, GPRCY4DRY-6, lymphocyte, 22 | |
| NS0 mouse myeloma cell line transfected with human CCR6, Met1-Met374, Accession # P51684 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction