Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR6 Antibody (MM0066-3L1), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212131AF700
Description
CCR6 Monoclonal specifically detects CCR6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CCR6 | |
Monoclonal | |
Alexa Fluor 700 | |
P51684 | |
CCR6 | |
Human CCR6 transfected 293 cells | |
0.1 mL | |
Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
1235 | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
MM0066-3L1 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
BN-1, CC-CKR-6, CCR-6, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29C-C CKR-6, GPRCY4DRY-6, lymphocyte, 22 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Detects human CCR6, not other CCRs | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction