Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR7 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
$215.16
Specifications
| Antigen | CCR7 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10, ELISA 1:100000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:250, Immunohistochemistry-Frozen 1:250, In vivo assay |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100712SS
![]() |
Novus Biologicals
NB100712SS |
0.02 mg |
Each of 1 for $215.16
|
|
|||||
Description
CCR7 Polyclonal specifically detects CCR7 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vivo assay.Specifications
| CCR7 | |
| Western Blot 1:1000, Flow Cytometry 1:10, ELISA 1:100000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:250, Immunohistochemistry-Frozen 1:250, In vivo assay | |
| Polyclonal | |
| Goat | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 10 mM KHPO4, 0.14 M NaCl, 1.0 mg/mL BSA with 0.1% Sodium Azide | |
| BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR-7, CD197, CD197 antigen, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7chemokine (C-C) receptor 7, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor | |
| CCR7 | |
| IgG | |
| Affinity Purified | |
| Peptide sequence is < 50 % identical to other mouse chemokine receptors in this region. |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| P47774 | |
| 1236 | |
| Synthetic peptide: DPGKPRKNVLVVALLVIFQVC, corresponding to amino acids 2-22 of Mouse CCR7. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 43 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title