Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR7 (CD197) Mouse anti-Human, Brilliant Violet 421, Clone: 2-L1-A, BD Horizon™
Mouse Monoclonal Antibody
$235.50 - $464.50
Specifications
| Antigen | CCR7 (CD197) |
|---|---|
| Clone | 2-L1-A |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 421 |
Description
CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow Cytometry.Specifications
| CCR7 (CD197) | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| RUO | |
| IgG1 κ | |
| Refrigerate (2 to 8°C) |
| 2-L1-A | |
| Monoclonal | |
| Mouse | |
| Human | |
| Primary | |
| CCR7 (CD197) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title