Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR7 (CD197) Mouse anti-Human , BV605, Clone: 2-L1-A, BD Biosciences™
Mouse Monoclonal Antibody
$225.00 - $444.00
Specifications
Antigen | CCR7 (CD197) |
---|---|
Clone | 2-L1-A |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 605 |
Description
CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow CytometrySpecifications
CCR7 (CD197) | |
Flow Cytometry | |
Brilliant Violet 605 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human CCR7 Transfected Cell Line | |
Primary |
2-L1-A | |
Monoclonal | |
Mouse | |
Human | |
BLR2; CC chemokine receptor 7; CMKBR7; EBI1; EVI1; Epstein-Barr virus induced gene 1; MIP-3 beta receptor | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV605 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV605 were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title