Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR7 (CD197) Mouse anti-Human, Clone: 2-L1-A, BD Pharmingen™

Catalog No. BDB567209 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB567209 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB567209 Supplier BD Biosciences Supplier No. 567209
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow Cytometry, Immunohistochemistry-frozen
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCR7 (CD197)
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 2-L1-A
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias BLR2; CC chemokine receptor 7; CMKBR7; EBI1; EVI1; Epstein-Barr virus induced gene 1; MIP-3 beta receptor
Host Species Mouse
Immunogen Human CCR7 Transfected Cell Line
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. Store undiluted at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.