Learn More
CCR7 (CD197) Mouse anti-Human , PE-CF594, Clone: 2-L1-A, BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CCR7 (CD197) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2-L1-A |
| Conjugate | PE-CF594 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | BLR2; CC chemokine receptor 7; CMKBR7; EBI1; EVI1; Epstein-Barr virus induced gene 1; MIP-3 beta receptor |
| Host Species | Mouse |
| Immunogen | Human CCR7 Transfected Cell Line |
| Quantity | 100 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.