Learn More
Description
Specifications
Specifications
| Antigen | CCR7 (CD197) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2-L1-A |
| Conjugate | PE |
| Host Species | Mouse |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Test Specificity | CCR7 (CD197) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.