Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR7 Rabbit anti-Mouse, Clone: 4B12, Novus Biologicals™

Rabbit Monoclonal Antibody
$280.00 - $596.50
Specifications
Antigen | CCR7 |
---|---|
Clone | 4B12 |
Concentration | 1 mg/ml |
Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional Assay |
Applications | Flow Cytometry, Immunofluorescence, Immunoprecipitation, Functional Assay |
Description
CCR7 Monoclonal antibody specifically detects CCR7 in Mouse samples. It is validated for Flow Cytometry, Immunofluorescence, Immunoprecipitation, Functional AssaySpecifications
CCR7 | |
1 mg/ml | |
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Functional Assay | |
Unconjugated | |
Rabbit | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
PBS | |
1236 | |
IgG κ | |
Protein A purified |
4B12 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional Assay | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR-7, CD197, CD197 antigen, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7chemokine (C-C) receptor 7, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor | |
This antibody was raised by immunizing rats with mouse CCR7 antigen. | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title