Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ccr7 Rat anti-Mouse, Clone:4B12, Abnova™

Catalog No. 89333058 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-058 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-058 Supplier Abnova Corporation Supplier No. MAB16951
Add to Cart
Add to Cart

Rat monoclonal antibody raised against mouse Ccr7.

Specifications

Antigen Ccr7
Applications Flow Cytometry
Classification Monoclonal
Clone 4B12
Conjugate Unconjugated
Description chemokine (C-C motif) receptor 7
Dilution Flow Cytometry
Immunoprecipitation
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (15 mM sodium azide)
Gene Ccr7
Gene Alias CD197, Cdw197, Cmkbr7, EBI1, Ebi1h
Host Species Rat
Immunogen Murine CD197-transfected RBL-2H3 cells.
Quantity 100 μg
Gene ID (Entrez) 12775
Test Specificity This antibody recognizes murine CD197 / CCR7 (chemokine receptor 7), a 43 kDa G-protein-coupled receptor for chemokines CCL19 and CCL21.
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.