Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR8 Antibody (6B12), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | CCR8 |
---|---|
Clone | 6B12 |
Dilution | Flow Cytometry |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CCR8 Monoclonal antibody specifically detects CCR8 in Human samples. It is validated for Flow CytometrySpecifications
CCR8 | |
Flow Cytometry | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR-8, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCY6, GPR-CY6, TER1CC chemokine receptor CHEMR1 | |
Recombinant Human CCR8 protein [UniProt P51685] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
6B12 | |
Flow Cytometry | |
Unconjugated | |
Human | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
1237 | |
IgG1 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title