Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR8 (CD198) Mouse anti-Human , APC, Clone: 433H, BD Biosciences™
Mouse Monoclonal Antibody
$164.70 - $367.50
Specifications
Antigen | CCR8 (CD198) |
---|---|
Clone | 433H |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC |
Description
CCR8 (CD198) Monoclonal antibody specifically detects CCR8 (CD198) in Human samples. It is validated for Flow CytometrySpecifications
CCR8 (CD198) | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human CCR8 Transfected Cell Line | |
Primary |
433H | |
Monoclonal | |
Mouse | |
Human | |
CCR8; CKR-L1; CKRL1; CMKBR8; CMKBRL2; CY6; GPR-CY6; GPRCY6; MGC129966; MGC1 | |
IgG2a κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to APC under optimum conditions, and unconjugated antibody and free APC were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title