Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR8 (CD198) Mouse anti-Human, R718, Clone: 433H, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 567766
This item is not returnable.
View return policy
Description
CCR8 (CD198) Monoclonal antibody specifically detects CCR8 (CD198) in Human samples. It is validated for Flow Cytometry
Specifications
CCR8 (CD198) | |
Monoclonal | |
R718 | |
CCR8; CKR-L1; CKRL1; CMKBR8; CMKBRL2; CY6; GPR-CY6; GPRCY6; MGC129966; MGC1 | |
Human CCR8 Transfected Cell Line | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
433H | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction