Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR8 Rabbit anti-Human, Mouse, Clone: 9N1Y1, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $504.00
Specifications
Antigen | CCR8 |
---|---|
Clone | 9N1Y1 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CCR8 Monoclonal antibody specifically detects CCR8 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
CCR8 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Virology Bacteria and Parasites | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
1237 | |
IgG | |
Affinity purified |
9N1Y1 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
CC chemokine receptor 8, C-C CKR-8, CC-chemokine receptor chemr1, CC-CKR-8, CCR-8, CDw198, CDw198 antigen, chemokine (C-C motif) receptor 8, chemokine (C-C) receptor 8, chemokine (C-C) receptor-like 2, Chemokine receptor-like 1, CKR-L1, CKRL1MGC129966, CMKBR8MGC129973, CMKBRL2C-C chemokine receptor type 8, CY6, GPRCY6, GPR-CY6, TER1CC chemokine receptor CHEMR1 | |
A synthetic peptide corresponding to a sequence within amino acids 250-355 of human CCR8 (P51685). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title