Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCR8 Recombinant Human Monoclonal Antibody (2C10)
GREENER_CHOICE

Catalog No. FEMA549966
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA549966 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA549966 Supplier Invitrogen™ Supplier No. MA549966
Add to Cart
Add to Cart

Human Recombinant Monoclonal Antibody

CCR8 Recombinant Monoclonal Antibody for Flow

CCR8 encodes a member of the beta chemokine receptor family, which is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptors are important for the migration of various cell types into the inflammatory sites. This receptor protein preferentially expresses in the thymus. I-309, thymus activation-regulated cytokine (TARC) and macrophage inflammatory protein-1 beta (MIP1 beta) have been identified as ligands of this receptor. Studies of this receptor and its ligands suggested its role in regulation of monocyte chemotaxis and thymic cell apoptosis. More specifically, this receptor may contribute to the proper positioning of activated T cells within the antigenic challenge sites and specialized areas of lymphoid tissues. This gene is located at the chemokine receptor gene cluster region. CCR8 expression has been reported in spleen and thymus, natural-killer cells, monocytes, and T cells, and at lower levels in peripheral blood leukocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCR8
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 2C10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene CCR8
Gene Accession No. P51685
Gene Alias C-C; CC chemokine receptor 8; CC chemokine receptor CHEMR1; C-C chemokine receptor type 8; C-C CKR-8; C-C motif chemokine receptor 8; CC-chemokine receptor chemr1; CC-CKR-8; CCR8; CCR-8; cd198; CDw198; chemokine (C-C motif) receptor 8; chemokine (C-C) receptor 8; chemokine (C-C) receptor-like 2; chemokine receptor-like 1; CKR-8; CKRL1; CKR-L1; Cmkbr8; CMKBRL2; CY6; GPRCY6; GPR-CY6; mCCR8; MGC129966; MGC129973; TER1
Gene Symbols CCR8
Host Species Human
Immunogen Recombinant Human CCR8 protein.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1237
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.