Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR9 Antibody (9B1), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP312001MFV610
Description
CCR9 Monoclonal antibody specifically detects CCR9 in Mouse samples. It is validated for Flow CytometrySpecifications
CCR9 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
9B1 | |
Flow Cytometry | |
C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR-9, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28 | |
This antibody was raised by immunizing rats with KLH-conjugated linear peptide corresponding to the N-terminus of mouse CCR9. | |
0.1 mL | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
10803 | |
Store at 4C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction