Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR9 Rabbit anti-Human, Mouse, Rat, Clone: 4F7Q6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | CCR9 |
---|---|
Clone | 4F7Q6 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CCR9 Monoclonal antibody specifically detects CCR9 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
CCR9 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
10803 | |
IgG | |
Affinity purified |
4F7Q6 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR-9, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CCR9 (P51686). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title