Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCR9 Recombinant Rabbit Monoclonal Antibody (4F7Q6)
GREENER_CHOICE

Catalog No. PIMA538201
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538201 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538201 Supplier Invitrogen™ Supplier No. MA538201
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: A-549, Jurkat, U-937, Mouse lung, Mouse spleen, Rat lung, Rat spleen Immunogen sequence: MTPTDFTSPI PNMADDYGSE STSSMEDYVN FNFTDFYCEK NNVRQFASHF LPPLYWLVFI VGALGNSLVI LVYWYCTRVK TMTDMFLLNL AIADLLFLVT.

The protein encoded by this gene is a member of the beta chemokine receptor family. It is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptors are key regulators of the thymocytes migration and maturation in normal and inflammation conditions. The specific ligand of this receptor is CCL25. It has been found that this gene is differentially expressed by T lymphocytes of small intestine and colon, suggested a role in the thymocytes recruitment and development that may permit functional specialization of immune responses in different segment of the gastrointestinal tract. This gene is mapped to the chemokine receptor gene cluster region. Two alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCR9
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 4F7Q6
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene Ccr9
Gene Accession No. P51686, Q9WUT7
Gene Alias A130091K22Rik; C-C chemokine receptor type 9; C-C CKR-9; C-C motif chemokine receptor 9; CC-CKR-9; Ccr9; CCR-9; CDw199; CDw199 antigen; chemokine (C-C motif) receptor 9; chemokine C-C receptor 10; Cmkbr10; G protein-coupled receptor 28; GPR28; GPR-9-6; G-protein coupled receptor 28
Gene Symbols Ccr9
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10803, 12769, 282832
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.