Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCRD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111711
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111711 100 μL
PIPA5110787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5111711 Supplier Invitrogen™ Supplier No. PA5111711
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QYLKAFLAAV LGWHLAPGTA QASLSSCSES SILTAQEEMT GMNDLGERQS ENYPNKEDVG NKSA Highest antigen sequence indentity to the following orthologs: Mouse - 56%, Rat - 56%.

This gene encodes a beta chemokine receptor, which is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptor-mediated signal transduction are critical for the recruitment of effector immune cells to the inflammation site. This gene is expressed in a range of tissues and hemopoietic cells. The expression of this receptor in lymphatic endothelial cells and overexpression in vascular tumors suggested its function in chemokine-driven recirculation of leukocytes and possible chemokine effects on the development and growth of vascular tumors. This receptor appears to bind the majority of beta-chemokine family members; however, its specific function remains unknown. This gene is mapped to chromosome 3p21.3, a region that includes a cluster of chemokine receptor genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCRD6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACKR2
Gene Accession No. O00590
Gene Alias Ackr2; AI464239; Atypical chemokine receptor 2; C-C chemokine receptor D6; Ccbp2; CC-chemokine-binding receptor JAB61; CCR10; CCR10-related receptor; CCR9; chemokine (C-C motif) receptor 9; chemokine (C-C) receptor 9; chemokine binding protein 2; chemokine decoy receptor D6; chemokine receptor CCR-10; Chemokine receptor CCR-9; chemokine receptor D6; chemokine-binding protein 2; Chemokine-binding protein D6; CMKBR9; D6; D6 beta-chemokine receptor; hD6
Gene Symbols ACKR2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human CCRD6. Recombinant protein control fragment (Product #RP-107935).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1238
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.