Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCRL1 Mouse anti-Human, BUV615, Clone: 13E11, BD OptiBuild™

Catalog No. BDB752303 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752303 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752303 Supplier BD Biosciences Supplier No. 752303
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCRL1 Monoclonal antibody specifically detects CCRL1 in Human samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CCRL1
Applications Flow Cytometry
Classification Monoclonal
Clone 13E11
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 615
Description Hu CCRL1 BUV615 13E11 50 μg
Host Species Mouse
Immunogen Human CCX-CKR gene-based immunization
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CCRL1
Target Species Human
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG2b κ
Reactivity Human
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.