Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCRL1 Mouse anti-Human, BV711, Clone: 13E11, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 747803
Description
CCRL1 Monoclonal antibody specifically detects CCRL1 in Human samples. It is validated for Flow Cytometry.
Specifications
CCRL1 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Human | |
Purified |
Flow Cytometry | |
13E11 | |
Brilliant Violet 711 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction