Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCRL2 Chimeric Recombinant Rabbit Monoclonal Antibody (BZ2E3)
GREENER_CHOICE

Catalog No. PIMA548143
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548143 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548143 Supplier Invitrogen™ Supplier No. MA548143
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: This antibody is specific for murine CCRL2 (formerly known as L-CCR), which is expressed on mast cells, activated macrophages, and endothelial cells. This receptor binds the leukocyte chemoattractant chemerin without triggering classical GPCR signaling, providing a specific mechanism for local enrichment of chemerin at inflammatory sites. CCRL2 presents active chemerin to leukocytes expressing the chemoattractant receptor CMKLR1/Chemr23. Chemerin can block anti-CCRL2 antibody binding. Preparation: This antibody was raised by immunising Wistar Furth rats with the mCCRL2 peptide/KLH conjugate. The immunizing amino-terminal mCCRL2 peptide with the sequence NH2-MDNYTVAPDDEYDVLILDDYLDNSC-COOH, corresponding to residues 1-24 of mCCRL2, with a nonnative carboxyl-terminal cysteine to facilitate conjugation to keyhole limpet hemocyanin (KLH).

CCRL2 (C-C chemokine receptor-like 2), also known as chemokine receptor X, human chemokine receptor (HCR), CRAM-A or CRAM-B, is a seven pass transmembrane protein expressed by monocytes, neutrophils, and dendritic cells of immunal tissues (predominanly spleen, fetal liver, bone marrow and lymph node). It shares over 40% homology with other C-C chemokine receptors but does not share the conserved DRY motif which is among the important motifs necessary for signalling and ligand-binding. C-C chemokine receptors are G-protein coupled, seven pass transmembrane domain proteins whose major physiological role is to function in the chemotaxis of T cells and phagocytic cells to areas of inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCRL2 Chimeric
Applications ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone BZ2E3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene CCRL2
Gene Accession No. O35457
Gene Alias 1810047I05Rik; Ackr5; atypical chemokine receptor 5; C-C chemokine receptor-like 2; C-C motif chemokine receptor like 2; CCR11; CCR6; Ccrl2; chemokine (C-C motif) receptor-like 2; chemokine C-C motif receptor-like 2; chemokine C-C motif receptor-like 2-like; chemokine receptor CCR11; Chemokine receptor X; CKRX; Cmkbr1l2; CRAM; CRAM-A; CRAM-B; E01; G-protein coupled beta chemokine receptor; HCR; Lccr; L-CCR; Lipopolysaccharide-inducible C-C chemokine receptor; LOC100719164; MGC116710; Putative MCP-1 chemokine receptor
Gene Symbols CCRL2
Host Species Rabbit
Immunogen This antibody was raised by immunising Wistar Furth rats with the mCCRL2 peptide (aa 1-24) /KLH conjugate.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54199
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.