Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCRL2/CRAM-A/B Antibody (152254), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB23501PEATT594
Description
CCRL2/CRAM-A/B Monoclonal antibody specifically detects CCRL2/CRAM-A/B in Human samples. It is validated for Flow CytometrySpecifications
CCRL2/CRAM-A/B | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
152254 | |
Flow Cytometry | |
C-C chemokine receptor-like 2, CCR11, CCR6, chemokine (C-C motif) receptor-like 2, Chemokine receptor CCR11, Chemokine receptor X, CKRXMGC116710, CRAM, CRAM-A, CRAM-B, FLJ55815, HCRMGC34104, Putative MCP-1 chemokine receptor | |
NS0 mouse myeloma cell line transfected with human CRAM-A, Met1-Val356 (Val180Met), Accession # NP_001124382 | |
0.5 mL | |
GPCR, Immunology, Innate Immunity | |
9034 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction