Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCRL2/CRAM-A/B Antibody (MM0158-5A15), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212137APC
Description
CCRL2/CRAM-A/B Monoclonal specifically detects CCRL2/CRAM-A/B in Human samples. It is validated for Flow Cytometry.Specifications
CCRL2/CRAM-A/B | |
Monoclonal | |
APC | |
NP_001124382 | |
CCRL2 | |
Human recombinant CCRL2/CRAM-A/B | |
0.1 mL | |
GPCR, Immunology, Innate Immunity | |
9034 | |
Human | |
Purified |
Flow Cytometry | |
MM0158-5A15 | |
Flow Cytometry | |
C-C chemokine receptor-like 2, CCR11, CCR6, chemokine (C-C motif) receptor-like 2, Chemokine receptor CCR11, Chemokine receptor X, CKRXMGC116710, CRAM, CRAM-A, CRAM-B, FLJ55815, HCRMGC34104, Putative MCP-1 chemokine receptor | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Detects both human CRAM-Aand CRAM-B isoforms. | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction