Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCRL2/CRAM-A/B Rabbit anti-Mouse, Clone: BZ5B8, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | CCRL2/CRAM-A/B |
---|---|
Clone | BZ5B8 |
Dilution | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional Assay |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CCRL2/CRAM-A/B Monoclonal antibody specifically detects CCRL2/CRAM-A/B in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Functional AssaySpecifications
CCRL2/CRAM-A/B | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional Assay | |
Unconjugated | |
RUO | |
Mouse | |
C-C chemokine receptor-like 2, CCR11, CCR6, chemokine (C-C motif) receptor-like 2, Chemokine receptor CCR11, Chemokine receptor X, CKRXMGC116710, CRAM, CRAM-A, CRAM-B, FLJ55815, HCRMGC34104, Putative MCP-1 chemokine receptor | |
This antibody was raised by immunising Wistar Furth rats with the mCCRL2/CRAM-A/B peptide/KLH conjugate. The immunizing amino-terminal mCCRL2/CRAM-A/B peptide with the sequence NH2-MDNYTVAPDDEYDVLILDDYLDNSC-COOH, corresponding to residues 1-24 of mCCRL2/CRAM-A/B, with a nonnative carboxyl-terminal cysteine to facilitate conjugation to keyhole limpet hemocyanin (KLH). | |
Primary |
BZ5B8 | |
Monoclonal | |
Rabbit | |
GPCR, Immunology, Innate Immunity | |
PBS | |
9034 | |
IgG κ | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title