Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCRL2/LCCR Antibody (498321) [FITC], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB5519F
Description
CCRL2/LCCR Monoclonal antibody specifically detects CCRL2/LCCR in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
CCRL2/LCCR | |
Monoclonal | |
FITC | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9034 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, Immunocytochemistry, CyTOF | |
498321 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
ACKR5;C-C chemokine receptor-like 2;CKRX;CRAM;CRAM-A;CRAM-B;HCR | |
RBL-1 rat basophilic leukemia cell line transfected with mouse CCRL2/LCCR, Accession # O35457 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction