Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCRL2 Monoclonal Antibody (498321)

Rat Monoclonal Antibody

Supplier:  Invitrogen™ MA524209

Catalog No. PIMA524209


Only null left
Add to Cart

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

CCRL2 (C-C chemokine receptor-like 2), also known as chemokine receptor X, human chemokine receptor (HCR), CRAM-A or CRAM-B, is a seven pass transmembrane protein expressed by monocytes, neutrophils, and dendritic cells of immunal tissues (predominanly spleen, fetal liver, bone marrow and lymph node). It shares over 40% homology with other C-C chemokine receptors but does not share the conserved DRY motif which is among the important motifs necessary for signalling and ligand-binding. C-C chemokine receptors are G-protein coupled, seven pass transmembrane domain proteins whose major physiological role is to function in the chemotaxis of T cells and phagocytic cells to areas of inflammation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CCRL2
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
O35457
CCRL2
RBL-1 rat basophilic leukemia cell line transfected with mouse CCRL2/LCCR.
100 μg
Primary
Mouse
Antibody
IgG1
Flow Cytometry, Immunocytochemistry
498321
Unconjugated
CCRL2
1810047I05Rik; Ackr5; atypical chemokine receptor 5; C-C chemokine receptor-like 2; C-C motif chemokine receptor like 2; CCR11; CCR6; Ccrl2; chemokine (C-C motif) receptor-like 2; chemokine C-C motif receptor-like 2; chemokine C-C motif receptor-like 2-like; chemokine receptor CCR11; Chemokine receptor X; CKRX; Cmkbr1l2; CRAM; CRAM-A; CRAM-B; E01; G-protein coupled beta chemokine receptor; HCR; Lccr; L-CCR; Lipopolysaccharide-inducible C-C chemokine receptor; LOC100719164; MGC116710; Putative MCP-1 chemokine receptor
Rat
Protein A/G
RUO
54199
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.