Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCRL2 (Mouse) Rat anti-Mouse, Clone: BZ5B8, MilliporeSigma™

Catalog No. MABF921MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF921MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF921MI Supplier MilliporeSigma Supplier No. MABF921
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This rat monoclonal Anti-CCRL2 (Mouse) Antibody, clone BZ5B8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCRL2 (Mouse)
Applications Flow Cytometry
Classification Monoclonal
Clone BZ5B8
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Ccrl2,Ccr11,Lccr
Host Species Rat
Immunogen KLH-conjugated linear peptide corresponding to the N-terminal end sequence of mouse CCRL2 (Zabel, B.A., et al. (2008). J. Exp. Med. 205(10):2207-2220).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001289305.1
Test Specificity Clone BZ5B8 reacts with mCCRL2, but not mCMKLR1, huCMKLR1, mCRTH2, huCCRL2, or mCCR10. Clone BZ5B8 immunostained the surface of mCCRL2-expressing cells, but not non-mCCRL2-expressing cells (Zabel, B.A., et al. (2008). J. Exp. Med. 205(10):2207-2220).
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.