Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCRL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575492
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575492 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575492 Supplier Invitrogen™ Supplier No. PA575492
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

CCRL2 (C-C chemokine receptor-like 2), also known as chemokine receptor X, human chemokine receptor (HCR), CRAM-A or CRAM-B, is a seven pass transmembrane protein expressed by monocytes, neutrophils, and dendritic cells of immunal tissues (predominanly spleen, fetal liver, bone marrow and lymph node). It shares over 40% homology with other C-C chemokine receptors but does not share the conserved DRY motif which is among the important motifs necessary for signalling and ligand-binding. C-C chemokine receptors are G-protein coupled, seven pass transmembrane domain proteins whose major physiological role is to function in the chemotaxis of T cells and phagocytic cells to areas of inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCRL2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CCRL2
Gene Accession No. O00421, O35457
Gene Alias 1810047I05Rik; Ackr5; atypical chemokine receptor 5; C-C chemokine receptor-like 2; C-C motif chemokine receptor like 2; CCR11; CCR6; Ccrl2; chemokine (C-C motif) receptor-like 2; chemokine C-C motif receptor-like 2; chemokine C-C motif receptor-like 2-like; chemokine receptor CCR11; Chemokine receptor X; CKRX; Cmkbr1l2; CRAM; CRAM-A; CRAM-B; E01; G-protein coupled beta chemokine receptor; HCR; Lccr; L-CCR; Lipopolysaccharide-inducible C-C chemokine receptor; LOC100719164; MGC116710; Putative MCP-1 chemokine receptor
Gene Symbols CCRL2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 191-236 of Human CCRL2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 316019, 54199, 9034
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.