Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCRN4L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552922
Description
Immunogen sequence: AVTHLKARTG WERFRSAQGC DLLQNLQNIT QGAKIPLIVC GDFNAEPTEE VYKHFASSSL NLNSAYKLLS ADGQSEPPYT TWKIRTSGEC RHTLDYIWYS KHALNVRSAL DLLTEEQIGP NRLPSFNYPS DHLSLVCDFS FT Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
CCRN4L is highly similar to Nocturnin, identified as a circadian clock regulated gene in Xenopus laevis. This protein and Nocturnin protein share similarity with the C-terminal domain of a yeast transcription factor, carbon catabolite repression 4 (CCR4).
Specifications
CCRN4L | |
Polyclonal | |
Unconjugated | |
NOCT | |
AU043840; carbon catabolite epression 4 homolog; carbon catabolite repression 4 homolog; carbon catabolite repression 4-like protein; CCR4; CCR4 carbon catabolite repression 4-like; CCR4 carbon catabolite repression 4-like (S. cerevisiae); CCR4 carbon catabolite repression 4-like B; CCR4 protein homolog; Ccr4c; CCR4L; CCR4-like (carbon catabolite repression 4, S.cerevisiae); CCRN4L; Ccrn4lb; circadian deadenylase NOC; MGC142054; MGC142060; MGC4120817; MGC78549; NOC; Noct; nocturin; nocturnin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25819 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UK39 | |
NOCT | |
Recombinant protein corresponding to Human CCRN4L. Recombinant protein control fragment (Product #RP-102539). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction