Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCRN4L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562830
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562830 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562830 Supplier Invitrogen™ Supplier No. PA562830
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ARSCSRTVCS MGTGTSRLYS ALAKTLNSSA ASQHPEYLVS PDPEHLEPID PKELLEECRA VLHTRPPRFQ RDFVDLRTDC PSTHPPIRVM QW Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

CCRN4L is highly similar to Nocturnin, identified as a circadian clock regulated gene in Xenopus laevis. This protein and Nocturnin protein share similarity with the C-terminal domain of a yeast transcription factor, carbon catabolite repression 4 (CCR4).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCRN4L
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NOCT
Gene Accession No. Q9UK39
Gene Alias AU043840; carbon catabolite epression 4 homolog; carbon catabolite repression 4 homolog; carbon catabolite repression 4-like protein; CCR4; CCR4 carbon catabolite repression 4-like; CCR4 carbon catabolite repression 4-like (S. cerevisiae); CCR4 carbon catabolite repression 4-like B; CCR4 protein homolog; Ccr4c; CCR4L; CCR4-like (carbon catabolite repression 4, S.cerevisiae); CCRN4L; Ccrn4lb; circadian deadenylase NOC; MGC142054; MGC142060; MGC4120817; MGC78549; NOC; Noct; nocturin; nocturnin
Gene Symbols NOCT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCRN4L. Recombinant protein control fragment (Product #RP-103168).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25819
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.