Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCS, Mouse anti-Human, Clone: 1G1, Abnova™

Catalog No. 89998905 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-905 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-905 Supplier Abnova Corporation Supplier No. MAB16178
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen CCS
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1G1
Conjugate Unconjugated
Dilution ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) Western Blot (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In HEPES, 150 mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide).
Gene copper chaperone for superoxide dismutase
Gene Alias MGC138260
Gene Symbols CCS
Host Species Mouse
Immunogen Recombinant protein corresponding to human CCS from E. coli.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9973
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.