Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCSER2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557967
Description
Immunogen sequence: IQLLELQLAT QHICHQKCKE EKCTYADKYT QTPWRRIPGG YSAPSFSPWQ GSFQGIPRTV PPHRRQTSST TAFQQPSQTH RSHPGKTNKA TT Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 60%.

Specifications
CCSER2 | |
Polyclonal | |
Unconjugated | |
CCSER2 | |
1700012P13Rik; 2900054P12Rik; AI035535; AW108503; bA486O22.1; Ccser2; coiled-coil serine rich 2; coiled-coil serine rich protein 2; coiled-coil serine-rich protein 2; FAM190B; family with sequence similarity 190, member B; Gcap14; granule cell antiserum positive 14; granule cell antiserum positive protein 14; KIAA1128; NPD012; protein FAM190B; Protein GCAP14; Protein GCAP14 homolog; RGD1304626; serine-rich coiled-coil domain-containing protein 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54462 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H7U1 | |
CCSER2 | |
Recombinant protein corresponding to Human CCSER2. Recombinant protein control fragment (Product #RP-97048). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction